missing translation for 'onlineSavingsMsg'
Learn More

R-96544 Hydrochloride, TRC

Product Code. 30402677
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402677 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30402677 Supplier Toronto Research Chemicals Supplier No. TRCH94292810MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material R-96544 Hydrochloride
CAS 167144-80-1
Molecular Formula C22H29NO3 . (HCl)
Quantity 10 mg
Synonym (3R,5R)-5-[2-[2-[2-(3-Methoxyphenyl)ethyl]phenoxy]ethyl]-1-methyl-3-pyrrolidinol Hydrochloride, (3R-trans)-5-[2-[2-[2-(3-Methoxyphenyl)ethyl]phenoxy]ethyl]-1-methyl-3-pyrrolidinol Hydrochloride, R 96544
InChI Formula InChI=1S/C22H29NO3.ClH/c1-23-16-20(24)15-19(23)12-13-26-22-9-4-3-7-18(22)11-10-17-6-5-8-21(14-17)25-2;/h3-9,14,19-20,24H,10-13,15-16H2,1-2H3;1H/t19-,20-;/m1./s1
SMILES O[C@H]1CN(C)[C@H](CCOC2=CC=CC=C2CCC3=CC=CC(OC)=C3)C1.Cl
IUPAC Name (3R,5R)-5-(2-(2-(3-methoxyphenethyl)phenoxy)ethyl)-1-methylpyrrolidin-3-ol hydrochloride
Molecular Weight (g/mol) 391.93
Formula Weight 391.67
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.