missing translation for 'onlineSavingsMsg'
Learn More

(R)-9-[2-(Diethylphosphonomethoxy)propyl] Adenine, TRC

Product Code. 30387240
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387240 10 mg -
30370640 5 mg 5mg
30378225 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387240 Supplier Toronto Research Chemicals Supplier No. TRCD44475010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 180587-75-1
Molecular Formula C13 H22 N5 O4 P
Molecular Weight (g/mol) 343.32
Synonym Phosphonic acid, P-[[(1R)-2-(6-amino-9H-purin-9-yl)-1-methylethoxy]methyl]-, diethyl ester, Phosphonic acid, [[(1R)-2-(6-amino-9H-purin-9-yl)-1-methylethoxy]methyl]-, diethyl ester (9CI), Phosphonic acid, [[2-(6-amino-9H-purin-9-yl)-1-methylethoxy]methyl]-, diethyl ester, (R)-, Diethyl (2R)-[[1-(6-Amino-9H-purin-9-yl)propan-2-yloxy]methyl]phosphonate, Diethyl [[(2R)-1-(6-Amino-9H-purin-9-yl)propan-2-yloxy]methyl]phosphonate
IUPAC Name 9-[(2R)-2-(diethoxyphosphorylmethoxy)propyl]purin-6-amine
SMILES CCOP(=O)(CO[C@H](C)Cn1cnc2c(N)ncnc12)OCC

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C13H22N5O4P/c1-4-21-23(19,22-5-2)9-20-10(3)6-18-8-17-11-12(14)15-7-16-13(11)18/h7-8,10H,4-6,9H2,1-3H3,(H2,14,15,16)/t10-/m1/s1
Formula Weight 343.1409
Chemical Name or Material (R)-9-[2-(Diethylphosphonomethoxy)propyl] Adenine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.