missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-8-Hydroxy-DPAT hydrobromide, R&D Systems™

Product Code. 30001363
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001363 50 mg 50mg
30001427 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001363 Supplier Tocris Bioscience Supplier No. 1080/50
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective 5-HT1A agonist; enantiomer of 8-Hydroxy-DPAT hydrobromide (Cat. No. 0529)

Full 5-HT1A serotonin receptor agonist. Reduces hippocampal 5-HT levels following systemic administration in rats in vivo. More active enantiomer of 8-Hydroxy-DPAT hydrobromide .

Racemate and 7-Hydroxy Isomer also available.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 78095-19-9
Molecular Formula C16H26BrNO
Molecular Weight (g/mol) 328.29
MDL Number MFCD00153810
InChI Key BATPBOZTBNNDLN-UHFFFAOYNA-N
Synonym r-+-8-hydroxy-dpat hydrobromide, unii-0s6ynt39th, 0s6ynt39th, r +-8-hydroxy dpat hbr, r-+-8-hydroxy-2-dipropylamino tetralin hydrobromide, r-+-2-dipropylamino-8-hydroxy-1,2,3,4-tetrahydronaphthalene hydrobromide, + 8-oh-dpat hydrobromide, c16h25no.hbr, 1-naphthalenol, 7-dipropylamino-5,6,7,8-tetrahydro-, hydrobromide, r
PubChem CID 11957570
IUPAC Name 7-(dipropylamino)-5,6,7,8-tetrahydronaphthalen-1-ol hydrobromide
SMILES Br.CCCN(CCC)C1CCC2=C(C1)C(O)=CC=C2

Tekniske data

Quantity 50 mg
Formula Weight 328.29
Percent Purity >99%
Product Line Tocris
Chemical Name or Material (R)-(+)-8-Hydroxy-DPAT hydrobromide
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.