missing translation for 'onlineSavingsMsg'
Learn More

(R)-8-(Diphenylphosphino)-1,2,3,4-tetrahydronaphthalen-1-amine, 97%, Thermo Scientific™

Product Code. 11995931 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11995931 250 mg Glass bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11995931 Supplier Thermo Scientific Acros Supplier No. 434112500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 960128-64-7
CAS Min % 96.0
CAS Max % 100.0
Melting Point 120.0°C to 127.0°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C22H22NP
MDL Number MFCD16621442
Quantity 250 mg
Synonym r-8-diphenylphosphino-1,2,3,4-tetrahydronaphthalen-1-amine, r-1-amino-8-diphenylphosphino-1,2,3,4-tetrahydronaphthalene, 1r-8-diphenylphosphanyl-1,2,3,4-tetrahydronaphthalen-1-amine, 1r-8-diphenylphosphino tetralin-1-amine, 1r-8-diphenylphosphanyl-1,2,3,4-tetrahydronaphthalen-1-ami, r-8-diphenylphosphanyl-1,2,3,4-tetrahydronaphthalen-1-amine, r---8-diphenylphosphino-1,2,3,4-tetrahydro-1-naphthylamine, r---8-diphenylphosphino-1,2,3,4-tetrahydro-1-naphthylamine, 97+%
InChI Key KTWOFTBMHDQUEE-GNLPSFAGNA-N
SMILES N[C@@H]1CCCC2=C1C(=CC=C2)P(C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name (1R)-8-diphenylphosphanyl-1,2,3,4-tetrahydronaphthalen-1-amine
Molecular Weight (g/mol) 331.40
PubChem CID 71309688
Formula Weight 331.39
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material (R)-8-(Diphenylphosphino)-1,2,3,4-tetrahydronaphthalen-1-amine
Show More Show Less
compliance-icons
Product Identifier
  • (R)-8-(Diphenylphosphino)-1,2,3,4-tetrahydronaphthalen-1-amine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.