Learn More
Tocris Bioscience™ R 59-022
Diacylglycerol kinase inhibitor; increases PKC activity
Specifications
Purity >98%
View More Specs
Description
Description
Diacylglycerol (DAG) kinase inhibitor (IC50 = 2.8 μM); increases protein kinase C activity. Potentiates thrombin-induced platelet aggregation and induces neutrophil chemotaxis. Inhibits U46619-induced contractions in mouse aorta and porcine coronary artery.
Specifications
Specifications
>98%