Learn More
IKA R 402 Stirring Shaft Protection
Product Code. 31910313 Shop All IKA Products
Change view
Click to view available options
Material:
Plastic
Unit Size:
Each
Product Code. Material Unit Size
31910313 Plastic Each
1 options
Product Code. 31910313 Supplier IKA Supplier No. 0020115312
  • ONLINE EXCLUSIVE PRICE
    £169.00
    £154.00 / Pack of 1
    Save £15.00
    8% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Stirring shaft protection for overhead stirrers to prevent users from potential injuries at the rotating shafts and stirring elements.
  • Equipped with a hinge to facilitate access to the stirrer chuck without having to remove it from the motor housing.
  • Can be attached directly to the overhead stirrers RW 20 digital, RW 28 digital, and the devices of the EUROSTAR series.
TRUSTED_SUSTAINABILITY
Specifications
Type Chuck Guard
Material Plastic