missing translation for 'onlineSavingsMsg'
Learn More

(R)-4-Hydroxy-6-(4-methoxyphenoxy)-4-methylhexanenitrile, TRC

Product Code. 30393732
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393732 10 mg 10mg
30392035 25 mg 25mg
30393740 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393732 Supplier Toronto Research Chemicals Supplier No. TRCH97280310MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (R)-4-Hydroxy-6-(4-methoxyphenoxy)-4-methylhexanenitrile
Analyte or Component Names (R)-4-Hydroxy-6-(4-methoxyphenoxy)-4-methylhexanenitrile
Shipping Condition Room Temperature
Molecular Formula C26 H21 F3 O3
InChI Formula InChI=1S/C26H21F3O3/c1-2-4-21(16-25(30)31)20-9-13-24(14-10-20)32-17-18-5-3-6-22(15-18)19-7-11-23(12-8-19)26(27,28)29/h3,5-15,21H,16-17H2,1H3,(H,30,31)/t21-/m1/s1
Solution Type Neat
SMILES CC#C[C@H](CC(=O)O)c1ccc(OCc2cccc(c2)c3ccc(cc3)C(F)(F)F)cc1
IUPAC Name (3R)-3-[4-[[3-[4-(trifluoromethyl)phenyl]phenyl]methoxy]phenyl]hex-4-ynoic acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 438.438
Quantity 10 mg
Formula Weight 438.144
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.