missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-4-Fluorostyrene oxide, 98+%, ee 98+%, Thermo Scientific Chemicals

Product Code. 15496447
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15496447 5 g 5g
15486447 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15496447 Supplier Thermo Scientific Alfa Aesar Supplier No. H33603.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134356-73-3
Molecular Formula C8H7FO
Molecular Weight (g/mol) 138.14
MDL Number MFCD03788740,MFCD00043994,MFCD03788740
InChI Key ICVNPQMUUHPPOK-UHFFFAOYNA-N
Synonym r-4-fluorophenyl oxirane, r-2-4-fluorophenyl oxirane, 2r-2-4-fluorophenyl oxirane, r-4-fluorostyrene oxide, r---4-fluorostyrene oxide, pubchem18166, r-4-fluorophenyl oxirane, technical gc
PubChem CID 7023123
IUPAC Name (2R)-2-(4-fluorophenyl)oxirane
SMILES FC1=CC=C(C=C1)C1CO1

Specifications

Density 1.17
Boiling Point 92°C (14 mmHg)
Refractive Index 1.5087
Quantity 5 g
UN Number UN1992
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
Formula Weight 138.14
Percent Purity ≥98%
Assay ee ≥98%
Chemical Name or Material (R)-(-)-4-Fluorostyrene oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.