missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-4-(Chloromethyl)-2,2-dimethyl-1,3-dioxolane, 98%, Thermo Scientific™

Product Code. 10234624 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10234624 1 g 1g
10612255 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10234624 Supplier Thermo Scientific Acros Supplier No. 399220010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 57044-24-3
Molecular Formula C6H11ClO2
Molecular Weight (g/mol) 150.60
MDL Number MFCD00273371
InChI Key BNPOTXLWPZOESZ-UHFFFAOYNA-N
Synonym r-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, r-+-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, 4r-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, 1,3-dioxolane, 4-chloromethyl-2,2-dimethyl-, 4r, r-3-chloro-1,2-propanediol acetonide, pubchem14074, ksc911s2r
PubChem CID 2734443
IUPAC Name (4R)-4-(chloromethyl)-2,2-dimethyl-1,3-dioxolane
SMILES CC1(C)OCC(CCl)O1

Specifications

Density 1.1030g/mL
Boiling Point 63°C (37.0 mmHg)
Flash Point 51°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Specific Gravity 1.103
Formula Weight 150.6
Percent Purity 98%
Chemical Name or Material (R)-(+)-4-(Chloromethyl)-2, 2-dimethyl-1, 3-dioxolane
compliance-icons compliance-icons compliance-icons
Product Identifier
  • (R)-(+)-4-(Chloromethyl)-2,2-dimethyl-1,3-dioxolane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H301-Toxic if swallowed.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.