missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-4-Benzyl-2-oxazolidinone, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | (R)-4-Benzyl-2-oxazolidinone |
| CAS | 102029-44-7 |
| Molecular Formula | C10 H11 N O2 |
| Quantity | 1 g |
| Synonym | 2-Oxazolidinone, 4-(phenylmethyl)-, (4R)-, 2-Oxazolidinone, 4-(phenylmethyl)-, (R)-, (4R)-4-(Phenylmethyl)-2-oxazolidinone, (+)-4-Benzyl-2-oxazolidinone, (4R)-4-(Phenylmethyl)-1,3-oxazolidin-2-one, (4R)-4-Benzyl-1,3-oxazolidin-2-one, (4R)-4-Benzyl-2-oxazolidinone, (R)-(+)-4-Benzyl-2-oxazolidinone, (R)-(+)-4-Benzyl-2-oxazolidinone, (R)-(+)-4-Benzyl-2-oxazolidone, (R)-4-(Phenylmethyl)-2-oxazolidinone, (R)-4-Benzyl-1,3-oxazolidin-2-one, (R)-4-Benzyl-2-oxazolidinone, (R)-4-Benzyl-2-oxooxazolidine, (R)-4-Benzyloxazolidin-2-one, 4(R)-Benzyloxazolidin-2-one |
| InChI Formula | InChI=1S/C10H11NO2/c12-10-11-9(7-13-10)6-8-4-2-1-3-5-8/h1-5,9H,6-7H2,(H,11,12)/t9-/m1/s1 |
| SMILES | O=C1N[C@H](Cc2ccccc2)CO1 |
| IUPAC Name | (4R)-4-benzyl-1,3-oxazolidin-2-one |
| Molecular Weight (g/mol) | 177.2 |
| Formula Weight | 177.079 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction