missing translation for 'onlineSavingsMsg'
Learn More

(R)-4-(1-Hydroxyethyl)pyridine, 99+%, (99+% ee), Thermo Scientific™

Product Code. 10116753 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10116753 250 mg Glass bottle 250mg
10208993 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10116753 Supplier Thermo Scientific Acros Supplier No. 393362500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 27854-88-2
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.15
MDL Number MFCD00077865
InChI Key HVOAMIOKNARIMR-ZCFIWIBFSA-N
Synonym r-1-pyridin-4-yl ethanol, r-+-1-4-pyridyl ethanol, 1r-1-pyridin-4-yl ethan-1-ol, r-1-4-pyridyl ethanol, 1r-1-pyridin-4-yl ethanol, r-4-1-hydroxyethyl pyridine, 1r-1-pyridin-4-ylethanol, r-1-pyridin-4-yl-ethanol, 1r-1-4-pyridyl ethanol
PubChem CID 11804747
IUPAC Name (1R)-1-pyridin-4-ylethanol
SMILES CC(C1=CC=NC=C1)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 67°C to 70°C
Color White
Enantiomeric Excess 99.0% min.
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Quantity 250 mg
Specific Rotation Condition +58.50° (20°C c=1,chloroform)
Specific Rotation + 58.5
Formula Weight 123.15
Percent Purity 99+%
Physical Form Crystalline Powder or Needles
Chemical Name or Material (R)-4-(1-Hydroxyethyl)pyridine
Show More Show Less
compliance-icons
Product Identifier
  • (R)-4-(1-Hydroxyethyl)pyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.