missing translation for 'onlineSavingsMsg'
Learn More

(R)-4-(1-Aminoethyl)phenol, 97%, Thermo Scientific Chemicals

Product Code. 15439428
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134855-89-3
Molecular Formula C8H12BrNO
Molecular Weight (g/mol) 218.094
MDL Number MFCD03844647
InChI Key PZBBMKOZPQAHRA-RGMNGODLSA-N
Synonym s-4-1-amino-ethyl-phenol hydrobromid, s---1-4-hydroxyphenyl ethylamine hydrobromide
PubChem CID 49758827
IUPAC Name 4-[(1S)-1-aminoethyl]phenol;hydrobromide
SMILES CC(C1=CC=C(C=C1)O)N.Br

Specifications

Melting Point 30.0°C to 32.0°C
Color White
Density 0.9200g/mL
Boiling Point 81.0°C (8.0 mmHg)
Flash Point 91°C
Packaging Glass Bottle
Quantity 5 g
UN Number 3259
Formula Weight 137.18
Percent Purity 97%
Physical Form Solid
Chemical Name or Material (R)-4-(1-Aminoethyl)phenol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'