missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-((S)-5-(4-Fluorophenyl)-5-hydroxypentanoyl)-4-phenyloxazolidin-2-one, TRC

Product Code. 30390057
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30390057 10 mg
30394973 50 mg
30399530 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390057 Supplier Toronto Research Chemicals Supplier No. TRCF59772010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 404874-94-8
Molecular Formula C20H20FNO4
Molecular Weight (g/mol) 357.38
Synonym 3-[(5S)-(4-Fluorophenyl)-5-hydroxypentanoyl]-(4R)-phenyl-1,3-oxazolidin-2-one
IUPAC Name (R)-3-((S)-5-(4-fluorophenyl)-5-hydroxypentanoyl)-4-phenyloxazolidin-2-one
SMILES O=C(N1C(OC[C@H]1C2=CC=CC=C2)=O)CCC[C@H](O)C3=CC=C(F)C=C3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C20H20FNO4/c21-16-11-9-15(10-12-16)18(23)7-4-8-19(24)22-17(13-26-20(22)25)14-5-2-1-3-6-14/h1-3,5-6,9-12,17-18,23H,4,7-8,13H2/t17-,18-/m0/s1
Formula Weight 357.14
Chemical Name or Material (R)-3-((S)-5-(4-Fluorophenyl)-5-hydroxypentanoyl)-4-phenyloxazolidin-2-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.