missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-3-Piperidinamine dihydrochloride, 97%, Thermo Scientific™

Product Code. 10431250 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10431250 1 g 1g
10361970 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10431250 Supplier Thermo Scientific Acros Supplier No. 436410010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 334618-23-4
Molecular Formula C5H12N2·2ClH
Molecular Weight (g/mol) 173.09
InChI Key GGPNYXIOFZLNKW-ZJIMSODOSA-N
Synonym r-3-aminopiperidine dihydrochloride, r-piperidin-3-amine dihydrochloride, r---3-aminopiperidine dihydrochloride, r-3-piperidinamine dihydrochloride, 3r-piperidin-3-amine dihydrochloride, r-3-amino-piperidine dihydrochloride, r---3-aminopiperidine 2hcl, r-3-aminopiperidine 2hcl, r-3-amino piperidine dihcl
PubChem CID 16211333
IUPAC Name (3R)-piperidin-3-amine;dihydrochloride
SMILES C1CC(CNC1)N.Cl.Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 207.5°C to 211.5°C
Color Tan to White
Elemental Analysis 33.5 to 35.9% C, 15.5 to 16.8% N
Infrared Spectrum Authentic
Assay Percent Range 97%
Quantity 1 g
Specific Rotation -1° to -3° (20°C, 589nm) (c=1, water)
Formula Weight 173.09
Percent Purity 97%
Physical Form Powder
Chemical Name or Material (R)-(-)-3-Piperidinamine dihydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • (R)-(-)-3-Piperidinamine dihydrochloride, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.