missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-3-Methyl-2-butanol, 98+%

Product Code. 11383136
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11383136 250 mg 250mg
11373136 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11383136 Supplier Thermo Scientific Alfa Aesar Supplier No. L11008.MD
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1572-93-6 | C5H12O | 88.15 g/mol

(R)-(-)-3-Methyl-2-butanol, is used as a pharmaceutical intermediate. It is also used as a solvent and an intermediate in the manufacture of other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-3-Methyl-2-butanol, is used as a pharmaceutical intermediate. It is also used as a solvent and an intermediate in the manufacture of other chemicals.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1572-93-6
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00065949
InChI Key MXLMTQWGSQIYOW-RXMQYKEDSA-N
Synonym 2r-3-methylbutan-2-ol, r---3-methyl-2-butanol, 2-butanol, 3-methyl-, 2r, r-3-methyl-2-butanol, r-3-methylbutan-2-ol, 2r-3-methyl-2-butanol, r---isopropyl methyl carbinol, unii-2nk7o363q6 component, unii-93ff0f303r component
PubChem CID 638099
IUPAC Name (2R)-3-methylbutan-2-ol
SMILES CC(C)C(C)O

Specifications

Boiling Point 112°C
Flash Point 26°C (78°F)
Quantity 250 mg
UN Number UN1105
Beilstein 3535088
Optical Rotation −5° (Neat)
Formula Weight 88.15
Percent Purity ≥98%
Chemical Name or Material (R)-(-)-3-Methyl-2-butanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.