missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-Isopropyl-2,5-dimethoxy-3,6-dihydropyrazine, 98%

Product Code. 15492456
Change view
Click to view available options
Quantity:
1 mL
5 mL
Unit Size:
1mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15492456 1 mL 1mL
15482456 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15492456 Supplier Thermo Scientific Alfa Aesar Supplier No. H56077.XF
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 109838-85-9 | C9H16N2O2 | 184.239 g/mol

(R)-3-Isopropyl-2,5-dimethoxy-3,6-dihydropyrazineChiral auxiliary for the synthesis of α-amino acids. It is an inhibitor with antitumor activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-3-Isopropyl-2,5-dimethoxy-3,6-dihydropyrazineChiral auxiliary for the synthesis of α-amino acids. It is an inhibitor with antitumor activity.

Solubility
Soluble in Chloroform and Ethanol

Notes
Store at -20°C. Keep away from oxidizing agents. Hygroscopic. Keep under inert atmosphere.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109838-85-9
Molecular Formula C9H16N2O2
Molecular Weight (g/mol) 184.239
MDL Number MFCD00040565
InChI Key FCFWEOGTZZPCTO-MRVPVSSYSA-N
Synonym r-2,5-dihydro-3,6-dimethoxy-2-isopropylpyrazine, r-2-isopropyl-3,6-dimethoxy-2,5-dihydropyrazine, pyrazine, 2,5-dihydro-3,6-dimethoxy-2-1-methylethyl-, 2r, r-2,5-dimethoxy-3-isopropyl-3,6-dihydropyrazine, 2r-2,5-dihydro-3,6-dimethoxy-2-isopropylpyrazine, 2r-2-isopropyl-3,6-dimethoxy-2,5-dihydropyrazine, r-2,5-dihydro-3,6-dimethoxy-2-isopropylpiperazine, 2r-3,6-dimethoxy-2-propan-2-yl-2,5-dihydropyrazine, 2r---2,5-dihydro-3,6-dimethoxy-2-isopropylpyrazine
PubChem CID 736066
IUPAC Name (2R)-3,6-dimethoxy-2-propan-2-yl-2,5-dihydropyrazine
SMILES CC(C)C1C(=NCC(=N1)OC)OC

Specifications

Density 1.028
Flash Point 85°C (185°F)
Quantity 1 mL
Beilstein 3542790
Solubility Information Soluble in Chloroform and Ethanol
Optical Rotation −102° (c=1 in Ethanol)
Formula Weight 184.24
Percent Purity 98%
Chemical Name or Material (R)-3-Isopropyl-2,5-dimethoxy-3,6-dihydropyrazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.