missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-3-Hydroxypyrrolidine Hydrochloride, TRC

Product Code. 30404646
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30404646 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30404646 Supplier Toronto Research Chemicals Supplier No. TRCH95305025G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (R)-(-)-3-Hydroxypyrrolidine Hydrochloride
CAS 104706-47-0
Molecular Formula C4H10ClNO
Quantity 25 g
Synonym (3R)-3-Hydroxypyrrolidine Hydrochloride, (3R)-3-Pyrrolidinol Hydrochloride, (R)-(-)-3-Pyrrolidinol Hydrochloride, (R)-3-Hydroxypyrrolidine Hydrochloride, (R)-3-Pyrrolidinol Hydrochloride, (R)-Pyrrolidin-3-ol Hydrochloride, 3-(R)-Hydroxypyrrolidine Hydrochloride
InChI Formula InChI=1S/C4H9NO.ClH/c6-4-1-2-5-3-4;/h4-6H,1-3H2;1H/t4-;/m1./s1
SMILES Cl.O[C@@H]1CCNC1
IUPAC Name (3R)-pyrrolidin-3-ol;hydrochloride
Molecular Weight (g/mol) 123.58
Formula Weight 123.0451
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.