missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-3-Hydroxypiperidine hydrochloride, 98%, Thermo Scientific™

Product Code. 15493388
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15493388 5 g 5g
15452936 250 mg 250mg
15483388 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15493388 Supplier Thermo Scientific Alfa Aesar Supplier No. H56847.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 198976-43-1
Molecular Formula C5H12ClNO
Molecular Weight (g/mol) 137.607
MDL Number MFCD00192221
InChI Key VLECDMDGMKPUSK-NUBCRITNSA-N
Synonym r-3-hydroxypiperidine hydrochloride, r-piperidin-3-ol hydrochloride, 3r-piperidin-3-ol hydrochloride, r-+-3-hydroxypiperidine hydrochloride, r-3-hydroxypiperidine hydrocloride, r-3-hydroxypiperidine hcl, r-+-3-piperidinol hydrochloride, r-3-hydroxylpiperidine hydrochloride, 3r-3-piperidinol hydrochloride, r-3-piperidinol hydrochloride
PubChem CID 2759333
IUPAC Name (3R)-piperidin-3-ol;hydrochloride
SMILES C1CC(CNC1)O.Cl

Specifications

Quantity 5 g
Optical Rotation +8.6° (c=2 in Methanol)
Formula Weight 137.61
Percent Purity 98%
Chemical Name or Material (R)-(+)-3-Hydroxypiperidine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.