missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-Hydroxybutanoic acid, MedChemExpress

Product Code. 16405467
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16405467 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16405467 Supplier Medchem Express Supplier No. HYW051723
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress (R)-3-Hydroxybutanoic acid is a metabolite, and converted from acetoacetic acid catalyzed by 3-hydroxybutyrate dehydrogenase. (R)-3-Hydroxybutanoic acid has applications as a nutrition source and as a precursor for vitamins, antibiotics and pheromones.

(R)-3-Hydroxybutanoic acid is a metabolite, and converted from acetoacetic acid catalyzed by 3-hydroxybutyrate dehydrogenase. (R)-3-Hydroxybutanoic acid has applications as a nutrition source and as a precursor for vitamins, antibiotics and pheromones.

Specifications

Chemical Name or Material (R)-3-Hydroxybutanoic acid
Physical Form Solid-Liquid Mixture
CAS 625-72-9
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C4H8O3
For Use With (Application) Metabolism-sugar/lipid metabolism
Synonym (R)-(-)-3-Hydroxybutanoic acid (R)-3-Hydroxybutyric acid
Solubility Information H2O : 100 mg/mL (960.61 mM; Need ultrasonic)
SMILES C[C@@H](O)CC(O)=O
Molecular Weight (g/mol) 104.1
Formula Weight 104.1
Percent Purity 97.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.