missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-3-Hydroxy Myristic Acid Dicyclohexylammonium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | (R)-3-Hydroxy Myristic Acid Dicyclohexylammonium Salt |
| CAS | 76062-98-1 |
| Molecular Formula | C26H51NO3 |
| Quantity | 250 mg |
| Synonym | (R)-3-Hydroxytetradecanoic Acid N-cyclohexylcyclohexanamine Salt, D-3-Hydroxytetradecanoic Acid N-cyclohexylcyclohexanamine Salt |
| InChI Formula | InChI=1S/C14H28O3.C12H23N/c1-2-3-4-5-6-7-8-9-10-11-13(15)12-14(16)17;1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h13,15H,2-12H2,1H3,(H,16,17);11-13H,1-10H2/t13-;/m1./s1 |
| SMILES | CCCCCCCCCCC[C@H](CC(=O)O)O.C1CCC(CC1)NC2CCCCC2 |
| IUPAC Name | N-cyclohexylcyclohexanamine;(3R)-3-hydroxytetradecanoic acid |
| Molecular Weight (g/mol) | 425.69 |
| Formula Weight | 425.3869 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?