missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-3-Fluoropyrrolidine hydrochloride, 97%

Product Code. 11461540
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11461540 1 g 1g
15477635 250 mg 250mg
11471540 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11461540 Supplier Thermo Scientific Alfa Aesar Supplier No. H26947.03
 Restricted Item
Estimated Shipment: 23-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 136725-55-8 | C4H9ClFN | 125.571 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136725-55-8
Molecular Formula C4H9ClFN
Molecular Weight (g/mol) 125.571
MDL Number MFCD04038717
InChI Key LENYOXXELREKGZ-PGMHMLKASA-N
Synonym r-3-fluoropyrrolidine hydrochloride, r---3-fluoropyrrolidine hydrochloride, 3r-3-fluoropyrrolidine hydrochloride, r---3-fluoropyrrolidinehydrochloride, r-3-fluoro-pyrrolidine hcl, 3r---3-fluoropyrrolidine hydrochloride, pyrrolidine, 3-fluoro-, hydrochloride, 3r, snxdjhbrbbruuuqx@, r-3-fluoropyrrolidine hcl
PubChem CID 2759162
IUPAC Name (3R)-3-fluoropyrrolidine;hydrochloride
SMILES C1CNCC1F.Cl

Specifications

Melting Point 181°C to 184°C
Quantity 1 g
Formula Weight 125.57
Percent Purity 97%
Chemical Name or Material (R)-(-)-3-Fluoropyrrolidine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.