missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-3-(Dimethylamino)pyrrolidine dihydrochloride, 97%, Thermo Scientific™

Product Code. p-7032788
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15418925 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15418925 Supplier Thermo Scientific Alfa Aesar Supplier No. H34401.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 864448-61-3
Molecular Formula C6H16Cl2N2
Molecular Weight (g/mol) 187.108
MDL Number MFCD08704347
InChI Key LCPKWRSLMCUOOZ-QYCVXMPOSA-N
Synonym r-+-3-dimethylamino pyrrolidine dihydrochloride, r-dimethylpyrrolidin-3-yl-amine dihydrochloride, r-n,n-dimethylpyrrolidin-3-amine dihydrochloride, 3r-n,n-dimethylpyrrolidin-3-amine dihydrochloride, r-3-dimethylaminopyrrolidine 2hcl, 3r-+-3-dimethylamino pyrrolidine 2hcl, r-3-dimethylamino pyrrolidine dihydrochloride, r-n,n-dimethyl-3-pyrrolidinamine.2hcl, r-3-dimethylaminopyrrolidine dihydrochloride, dimethyl-pyrrolidin-3-yl-amine dihydrochloride
PubChem CID 53487164
IUPAC Name (3R)-N,N-dimethylpyrrolidin-3-amine;dihydrochloride
SMILES CN(C)C1CCNC1.Cl.Cl

Specifications

Melting Point 194°C to 197°C
Quantity 250 mg
Sensitivity Hygroscopic
Optical Rotation +3.1° (c=1 in Water)
Formula Weight 187.11
Percent Purity 97%
Chemical Name or Material (R)-(+)-3-(Dimethylamino)pyrrolidine dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.