missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-(Boc-amino)-3-(3-methoxyphenyl)propionic acid, 97%, Thermo Scientific™

Product Code. 15429495
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15429495 250 mg 250mg
15409495 1 g 1g
15419495 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15429495 Supplier Thermo Scientific Alfa Aesar Supplier No. H52001.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals, petrochemicals, and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 500788-86-3
Molecular Formula C15H21NO5
Molecular Weight (g/mol) 295.335
MDL Number MFCD03427937
InChI Key DNXONWHJJNOKEU-GFCCVEGCSA-N
Synonym r-3-tert-butoxycarbonyl amino-3-3-methoxyphenyl propanoic acid, r-boc-3-methoxy-beta-phe-oh, boc-r-3-amino-3-3-methoxy-phenyl-propionic acid, boc-r-3-amino-3-3-methoxyphenyl propionic acid, r-3-boc-amino-3-3-methoxyphenyl propionic acid, 3r-3-tert-butoxycarbonyl amino-3-3-methoxyphenyl propanoic acid, 3r-3-tert-butoxy carbonylamino-3-3-methoxyphenyl propanoic acid, pubchem24031, boc-d-?-phe 3-ome-oh, r-boc-3-methoxy-, a-phe-oh
PubChem CID 2761760
IUPAC Name (3R)-3-(3-methoxyphenyl)-3-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)O)C1=CC(=CC=C1)OC

Specifications

Melting Point 106°C to 108°C
Quantity 250 mg
Optical Rotation +40° (c=1 in Ethanol)
Formula Weight 295.34
Percent Purity 97%
Chemical Name or Material (R)-3-(Boc-amino)-3-(3-methoxyphenyl)propionic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.