missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-(Boc-amino)-3-(3-hydroxyphenyl)propionic acid, 95%, Thermo Scientific™

Product Code. 15429595
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15429595 250 mg 250mg
15409595 1 g 1g
15419595 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15429595 Supplier Thermo Scientific Alfa Aesar Supplier No. H52109.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 500788-89-6
Molecular Formula C14H19NO5
Molecular Weight (g/mol) 281.308
MDL Number MFCD03427940
InChI Key KLIMLFKCLBBZKH-LLVKDONJSA-N
Synonym boc-r-3-amino-3-3-hydroxy-phenyl-propionic acid, r-3-boc-amino-3-3-hydroxyphenyl propionic acid, r-3-tert-butoxycarbonyl amino-3-3-hydroxyphenyl propanoic acid, 3r-3-tert-butoxycarbonyl amino-3-3-hydroxyphenyl propanoic acid, benzenepropanoic acid, b-1,1-dimethylethoxy carbonyl amino-3-hydroxy-, br, pubchem24049, pubchem24051, boc-d-beta-m-tyr-oh, boc-d-?-phe 3-oh-oh, boc-d-beta-phe 3-oh-oh
PubChem CID 2761766
IUPAC Name (3R)-3-(3-hydroxyphenyl)-3-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)O)C1=CC(=CC=C1)O

Specifications

Quantity 250 mg
Formula Weight 281.31
Percent Purity 95%
Chemical Name or Material (R)-3-(Boc-amino)-3-(3-hydroxyphenyl)propionic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.