missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
(R)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid is used as catalyst or pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(R)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid is used as catalyst or pharmaceutical intermediates.
Solubility
Insoluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
(R)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid is used as catalyst or pharmaceutical intermediates.
Solubility
Insoluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
Quantity | 1 g |
Solubility Information | Insoluble in water. |
Formula Weight | 426.46 |
Percent Purity | 95% |
Physical Form | Solid |
Chemical Name or Material | (R)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'