missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde, 95%, Thermo Scientific Chemicals

Product Code. 15448266
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

(R)-(+)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde is used in the preparation of fluorinated analogs of glutemic and glutamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde is used in the preparation of fluorinated analogs of glutemic and glutamine.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95715-87-0
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.28
MDL Number MFCD00674064
InChI Key PNJXYVJNOCLJLJ-QMMMGPOBSA-N
Synonym r-tert-butyl 4-formyl-2,2-dimethyloxazolidine-3-carboxylate, unii-upk623lv60, r-4-formyl-2,2-dimethyl-oxazolidine-3-carboxylic acid tert-butyl ester, tert-butyl 4r-4-formyl-2,2-dimethyl-1,3-oxazolidine-3-carboxylate, r-+-3-boc-2,2-dimethyloxazolidine-4-carboxaldehyde, tert-butyl r-+-4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate, 3-oxazolidinecarboxylic acid, 4-formyl-2,2-dimethyl-, 1,1-dimethylethyl ester, 4r, r-garner aldehyde, r-3-boc-4-formyl-2,2-dimethyl-1,3-oxazolidine, r---3-boc-2,2-dimethyloxazolidine-4-carboxaldehyde
PubChem CID 178792
IUPAC Name tert-butyl (4R)-4-formyl-2,2-dimethyl-1,3-oxazolidine-3-carboxylate
SMILES CC(C)(C)OC(=O)N1[C@H](COC1(C)C)C=O

Specifications

Color Colorless to Yellow
Density 1.06 g/mL
Boiling Point 67°C (0.3 mmHg)
Flash Point 102°C (216°F)
Refractive Index 1.445
Quantity 250 mg
Beilstein 3651554
Sensitivity Air sensitive
Merck Index 14,4371
Formula Weight 229.27
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material (R)-(+)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'