missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-3-Aminopyrrolidine dihydrochloride, 97%, Thermo Scientific Chemicals

Product Code. 11347616
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11347616 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11347616 Supplier Thermo Scientific Alfa Aesar Supplier No. L11173.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116183-81-4
Molecular Formula C4H12Cl2N2
Molecular Weight (g/mol) 159.054
MDL Number MFCD00070604
InChI Key NJPNCMOUEXEGBL-RZFWHQLPSA-N
Synonym 3r---3-aminopyrrolidine dihydrochloride, r---3-aminopyrrolidine dihydrochloride, r-pyrrolidin-3-amine dihydrochloride, r-3-aminopyrrolidine 2hcl, r-+-3-aminopyrrolidine 2hcl, 3r-pyrrolidin-3-amine dihydrochloride, 3r-3-pyrrolidinamine dihydrochloride, r-3-aminopyrrolidine dihydrochloride, 3-pyrrolidinamine, dihydrochloride, r, pubchem5730
PubChem CID 13500643
IUPAC Name (3R)-pyrrolidin-3-amine;dihydrochloride
SMILES C1CNCC1N.Cl.Cl

Specifications

Melting Point ∽283°C (decomposition)
Quantity 250 mg
Beilstein 5852700
Sensitivity Hygroscopic
Optical Rotation −1.2° (c=10 in Water)
Formula Weight 159.06
Percent Purity 98%
Chemical Name or Material (R)-(-)-3-Aminopyrrolidine dihydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.