missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-Aminomethyl-1-Boc-piperidine, 97%

Product Code. 15449307
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15449307

Brand: Thermo Scientific Alfa Aesar H52561.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 140645-23-4 | C11H22N2O2 | 214.309 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140645-23-4
Molecular Formula C11H22N2O2
Molecular Weight (g/mol) 214.309
MDL Number MFCD03839876
InChI Key WPWXYQIMXTUMJB-SECBINFHSA-N
Synonym r-1-boc-3-aminomethyl piperidine, r-n-boc-3-aminomethylpiperidine, r-tert-butyl 3-aminomethyl piperidine-1-carboxylate, r-1-n-boc-3-aminomethyl piperidine, tert-butyl 3r-3-aminomethyl piperidine-1-carboxylate, r-3-aminomethyl-1-n-boc-piperidine, r-3-aminomethyl-1-boc-piperidine, r-3-aminomethyl-piperidine-1-carboxylic acid tert-butyl ester, 3r-3-aminomethyl-n-tert-butoxycarbonylpiperidine
PubChem CID 1502023
IUPAC Name tert-butyl (3R)-3-(aminomethyl)piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(C1)CN

Specifications

Quantity 1 g
Formula Weight 214.31
Percent Purity 97%
Chemical Name or Material (R)-3-Aminomethyl-1-Boc-piperidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.