missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-Amino-3-phenylpropionic acid, 95%

Product Code. 15428527
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15428527

Brand: Thermo Scientific Alfa Aesar H52034.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 83649-48-3 | C9H12ClNO2 | 201.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83649-48-3
Molecular Formula C9H12ClNO2
Molecular Weight (g/mol) 201.65
MDL Number MFCD01076237
InChI Key ABEBCTCOPRULFS-DDWIOCJRSA-N
Synonym r-3-amino-3-phenylpropanoic acid hydrochloride, r-3-amino-3-phenylpropionic acid hydrochloride, 3r-3-amino-3-phenylpropanoic acid hydrochloride, l-beta-homophenylglycine hydrochloride, r-3-amino-3-phenylpropionic acid hcl, pubchem5925, d-beta-phenylalanine hcl, r-beta-homophenylglycine hydrochloride, 3r-3-amino-3-phenylpropanoic acid, chloride, r-3-amino-3-phenyl-propionic acid hcl salt
PubChem CID 2734697
IUPAC Name (3R)-3-amino-3-phenylpropanoic acid;hydrochloride
SMILES C1=CC=C(C=C1)C(CC(=O)O)N.Cl

Specifications

Quantity 250 mg
Formula Weight 165.19
Sensitivity Air Sensitive
Percent Purity 95%
Chemical Name or Material (R)-3-Amino-3-phenylpropionic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.