missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-Amino-3-phenylpropanoic acid ethyl ester hydrochloride, 95%, 98% ee

Product Code. p-57642 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10551292 1 g Glass Bottle 1g
10582232 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10551292 Supplier Thermo Scientific Acros Supplier No. 362990010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 340188-50-3 | C11H15NO2·HCl | 229.7 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 340188-50-3
Molecular Formula C11H15NO2·HCl
Molecular Weight (g/mol) 229.7
MDL Number MFCD02259750
InChI Key ATSZQDTVNRNXKB-HNCPQSOCSA-N
Synonym r-3-amino-3-phenylpropanoic acid ethyl ester hydrochloride, r-ethyl 3-amino-3-phenylpropanoate hydrochloride, ethyl 3r-3-amino-3-phenylpropanoate hydrochloride, r-3-amino-3-phenylpropanoic acid ethyl ester hcl, 3r-3-amino-3-phenylpropanoic acid ethyl ester hcl, pubchem13851, r-beta-phenylalanine ethyl ester hydrochloride, r-3-amino-3-phenylpropanoic acid ethyl ester hyd, r-3-amino-3-phenylpropionic acid ethyl ester hcl, r-3-amino-3-phenyl-propionic acid ethyl ester hydro chloride
PubChem CID 2734508
IUPAC Name ethyl (3R)-3-amino-3-phenylpropanoate;hydrochloride
SMILES CCOC(=O)CC(C1=CC=CC=C1)N.Cl

Specifications

Melting Point 119°C
CAS Min % 94.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 229.7
Percent Purity 95%,98% ee
Physical Form Powder
Chemical Name or Material (R)-3-Amino-3-phenylpropanoic acid ethyl ester hydrochloride, 95%, (98% e.e.)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.