missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-Amino-3-phenylpropan-1-ol, TRC

Product Code. 30326472
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Product Code. Quantity unitSize
30326472 500 mg 500mg
1 options
This item is not returnable. View return policy

Product Code. 30326472

Brand: Toronto Research Chemicals TRCA615915500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (R)-3-Amino-3-phenylpropan-1-ol
CAS 170564-98-4
Molecular Formula C9 H13 N O
Quantity 500 mg
Synonym Maraviroc Impurity 13
InChI Formula InChI=1S/C9H13NO/c10-9(6-7-11)8-4-2-1-3-5-8/h1-5,9,11H,6-7,10H2/t9-/m1/s1
SMILES N[C@H](CCO)c1ccccc1
IUPAC Name (3R)-3-amino-3-phenylpropan-1-ol
Molecular Weight (g/mol) 151.21
Formula Weight 151.0997
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.