missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-Amino-3-phenylpropan-1-ol, 95%, 98% ee, Thermo Scientific™

Product Code. 10180802 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10180802 1 g Glass bottle 1g
10272712 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10180802 Supplier Thermo Scientific Acros Supplier No. 363120010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 170564-98-4
MDL Number MFCD01311791
InChI Key SEQXIQNPMQTBGN-SECBINFHSA-N
Synonym r-1-phenyl-3-propanolamine, r-3-amino-3-phenylpropan-1-ol, 3r-3-amino-3-phenylpropan-1-ol, r-beta-phenylalaninol, r-3-amino-3-phenyl-1-propanol, r-3-phenyl-beta-alaninol, r-3-amino-3-phenyl-propan-1-ol, r--phenylalaninol, pubchem13853
PubChem CID 7016858
IUPAC Name (3R)-3-amino-3-phenylpropan-1-ol
SMILES C1=CC=C(C=C1)C(CCO)N

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color White to Yellow or Yellow
Enantiomeric Excess 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Molecular Formula C9H13NO
Quantity 1 g
Molecular Weight (g/mol) 151.21
Formula Weight 151.21
Percent Purity 95%,98% ee
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material (R)-3-Amino-3-phenylpropan-1-ol, 95%, (98% e.e.)
Show More Show Less
compliance-icons
Product Identifier
  • (R)-3-Amino-3-phenylpropan-1-ol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.