missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-3-Amino-1-(benzyloxycarbonyl)pyrrolidine, 96%, Thermo Scientific™

Product Code. 15434456
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15434456

Brand: Thermo Scientific Alfa Aesar H58688.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122536-73-6
Molecular Formula C12H16N2O2
Molecular Weight (g/mol) 220.272
MDL Number MFCD05864624
InChI Key FPXJNSKAXZNWMQ-LLVKDONJSA-N
Synonym 3r-1-n-cbz-3-aminopyrrolidine, r-3-amino-1-n-cbz-pyrrolidine, benzyl 3r-3-aminopyrrolidine-1-carboxylate, r-cbz-3-aminopyrrolidine, r-benzyl 3-aminopyrrolidine-1-carboxylate, r---1-cbz-3-aminopyrrolidine, r-1-cbz-3-aminopyrrolidine, r-3-amino-1-cbz-pyrrolidine, r-1-n-cbz-3-aminopyrrolidine, r-1-benzyloxycarbonyl-3-aminopyrrolidine
PubChem CID 1519356
IUPAC Name benzyl (3R)-3-aminopyrrolidine-1-carboxylate
SMILES C1CN(CC1N)C(=O)OCC2=CC=CC=C2

Specifications

Melting Point 310°C to 316°C
Density 1.155
Flash Point >110°C (230°F)
Refractive Index 1.54
Quantity 250 mg
Sensitivity Air Sensitive
Optical Rotation −5° (c=1 in Methanol)
Formula Weight 220.27
Percent Purity 96%
Chemical Name or Material (R)-(-)-3-Amino-1-(benzyloxycarbonyl)pyrrolidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.