missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-(4-Phenoxyphenyl)-1-(piperidin-3-yl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine, TRC

Product Code. 17990485
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17990485 1 g 1g
17902295 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17990485 Supplier Toronto Research Chemicals Supplier No. TRCP3187701G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1022150-12-4
Molecular Formula C22 H22 N6 O
Molecular Weight (g/mol) 386.45
Synonym 1H-Pyrazolo[3,4-d]pyrimidin-4-amine, 3-(4-phenoxyphenyl)-1-(3R)-3-piperidinyl- (ACI), 3-(4-Phenoxyphenyl)-1-[(3R)-1-(phenylmethyl)-3-piperidinyl]-1H-pyrazolo[3,4-d]pyrimidin-4-amine (ACI), 3-(4-Phenoxyphenyl)-1-(3R)-3-piperidinyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine
IUPAC Name 3-(4-phenoxyphenyl)-1-[(3R)-piperidin-3-yl]pyrazolo[3,4-d]pyrimidin-4-amine
SMILES Nc1ncnc2c1c(nn2[C@@H]3CCCNC3)c4ccc(Oc5ccccc5)cc4

Specifications

Quantity 1 g
InChI Formula InChI=1S/C22H22N6O/c23-21-19-20(15-8-10-18(11-9-15)29-17-6-2-1-3-7-17)27-28(22(19)26-14-25-21)16-5-4-12-24-13-16/h1-3,6-11,14,16,24H,4-5,12-13H2,(H2,23,25,26)/t16-/m1/s1
Formula Weight 386.1855
Chemical Name or Material (R)-3-(4-Phenoxyphenyl)-1-(piperidin-3-yl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.