missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-(4-Fluorophenyl)pyrrolidine, TRC

Product Code. 30402265
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30402265 50 mg
30391555 250 mg
30396612 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30402265 Supplier Toronto Research Chemicals Supplier No. TRCF23655050MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1048703-18-9
Molecular Formula C10H12FN
Molecular Weight (g/mol) 165.21
Synonym (3R)-3-(4-Fluorophenyl)pyrrolidine
SMILES Fc1ccc(cc1)[C@H]2CCNC2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C10H12FN/c11-10-3-1-8(2-4-10)9-5-6-12-7-9/h1-4,9,12H,5-7H2/t9-/m0/s1
Formula Weight 165.0954
Chemical Name or Material (R)-3-(4-Fluorophenyl)pyrrolidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.