missing translation for 'onlineSavingsMsg'
Learn More

R-268712, MedChemExpress

Product Code. 16496036
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496036 100 mg 100mg
16478157 5 mg 5mg
16435448 10 mg 10mg
16431627 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16496036 Supplier Medchem Express Supplier No. HY12953

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress R-268712 is an orally active and selective ALK-5 inhibitor, with an IC50 of 2.5 nM. R-268712 inhibits the phosphorylation of Smad3 in a dose-dependent manner with an IC50 of 10.4 nM. R-268712 suppresses glomerulonephritis as well as glomerulosclerosis by inhibiting TGF-β signaling, which can be used in studies of renal fibrosis and cancer.

R-268712 is an orally active and selective ALK-5 inhibitor, with an IC50 of 2.5 nM. R-268712 inhibits the phosphorylation of Smad3 in a dose-dependent manner with an IC50 of 10.4 nM. R-268712 suppresses glomerulonephritis as well as glomerulosclerosis by inhibiting TGF-β signaling, which can be used in studies of renal fibrosis and cancer.

Specifications

Chemical Name or Material R-268712
CAS 879487-87-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H18FN5O
Quantity 100 mg
Solubility Information DMSO : 125 mg/mL (343.98 mM; Need ultrasonic)
SMILES FC1=C(C2=CN(CCO)N=C2)C=C(C3=CNN=C3C4=CC=CC(C)=N4)C=C1
Molecular Weight (g/mol) 363.39
Formula Weight 363.39
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.