missing translation for 'onlineSavingsMsg'
Learn More

(R)-2-(Trifluoromethyl)piperidine, 97%, Thermo Scientific Chemicals

Product Code. 11956481 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11956481 1 g 1g
11966481 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11956481 Supplier Thermo Scientific Acros Supplier No. 446690010
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1212734-83-2
Molecular Formula C6H10F3N
Molecular Weight (g/mol) 153.15
InChI Key NAXDEFXCCITWEU-RXMQYKEDSA-N
Synonym r-2-trifluoromethyl piperidine, 2r-2-trifluoromethyl piperidine, r-2-trifluoromethylpiperidine
PubChem CID 42580053
IUPAC Name (2R)-2-(trifluoromethyl)piperidine
SMILES C1CCNC(C1)C(F)(F)F

Specifications

Boiling Point 80.0°C (200.0 Torr)
Assay Percent Range 96% min. (GC)
Quantity 1 g
Formula Weight 153.15
Percent Purity 97%
Chemical Name or Material (R)-2-(Trifluoromethyl)piperidine
compliance-icons compliance-icons compliance-icons
Product Identifier
  • (R)-2-(Trifluoromethyl)piperidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.