missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Phenylbutyric Acid, TRC

Product Code. 17981525
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17981525 500 mg 500mg
17903235 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17981525 Supplier Toronto Research Chemicals Supplier No. TRCP319795500MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 938-79-4
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.2
Synonym (alphaR)-alpha-Ethylbenzeneacetic Acid, (-)-(R)-2-Phenylbutyric Acid, (-)-2-Phenylbutanoic Acid, (R)-(-)-alpha-Ethylphenylacetic Acid, (R)-alpha-Ethylbenzeneacetic cid
IUPAC Name (2R)-2-phenylbutanoic acid
SMILES CC[C@@H](C(=O)O)c1ccccc1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C10H12O2/c1-2-9(10(11)12)8-6-4-3-5-7-8/h3-7,9H,2H2,1H3,(H,11,12)/t9-/m1/s1
Formula Weight 164.0837
Chemical Name or Material (R)-(-)-2-Phenylbutyric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.