missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Octanol, 99%

Product Code. 11342457
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11342457 1 g 1g
11352457 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11342457 Supplier Thermo Scientific Alfa Aesar Supplier No. L11502.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5978-70-1 | C8H18O | 130.231 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5978-70-1
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.231
MDL Number MFCD00064284
InChI Key SJWFXCIHNDVPSH-MRVPVSSYSA-N
Synonym r-octan-2-ol, r---2-octanol, r-2-octanol, l-octan-2-ol, 2r-octan-2-ol, 2r-2-octanol, unii-2shb67xf6h, d-2-octanol, r---hexylmethylcarbinol, l--2-octanol
PubChem CID 80080
ChEBI CHEBI:37871
IUPAC Name (2R)-octan-2-ol
SMILES CCCCCCC(C)O

Specifications

Density 0.819
Boiling Point 179°C to 180°C
Flash Point 76°C (169°F)
Odor Pleasant
Refractive Index 1.426
Quantity 1 g
Beilstein 1719324
Merck Index 14,6752
Optical Rotation −10° (c=1 in Ethanol)
Formula Weight 130.23
Percent Purity 99%
Chemical Name or Material (R)-(-)-2-Octanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.