missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Methylpyrrolidine, 95%

Product Code. 15454278
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15454278 1 g 1g
15464278 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15454278 Supplier Thermo Scientific Alfa Aesar Supplier No. H57885.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 41720-98-3 | C5H11N | 85.15 g/mol

It is used as a building block used in a multikilogram synthesis of a chiral naphthalenoid histamine receptor antagonist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a building block used in a multikilogram synthesis of a chiral naphthalenoid histamine receptor antagonist.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Keep container in a cool refrigerated, well-ventilated area. Moisture sensitive
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41720-98-3
Molecular Formula C5H11N
Molecular Weight (g/mol) 85.15
MDL Number MFCD07783026
InChI Key RGHPCLZJAFCTIK-RXMQYKEDSA-N
Synonym r-2-methylpyrrolidine, r-2-methyl-pyrrolidine, 2r-2-methylpyrrolidine, r---2-methylpyrrolidine, r-2-methyl pyrrolidine, 2-r-methylpyrrolidine, 2r---2-methyltetrahydropyrrole, 2r---2-methylpyrrolidine, pyrrolidine, 2-methyl-, 2r, 2-methyl-pyrrolidine
PubChem CID 641544
ChEBI CHEBI:78858
IUPAC Name (2R)-2-methylpyrrolidine
SMILES CC1CCCN1

Specifications

Density 0.84
Flash Point 7°C (45°F)
Refractive Index 1.43
Quantity 1 g
UN Number UN2733
Solubility Information Insoluble in water.
Optical Rotation −19° (c=5 in Methanol)
Formula Weight 85.15
Percent Purity 95%
Chemical Name or Material (R)-(-)-2-Methylpyrrolidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.