missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Methylpiperazine, 95%

Product Code. 10109841 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10109841 5 g 5g
11916531 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10109841 Supplier Thermo Scientific Acros Supplier No. 356270050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 75336-86-6 | C5H12N2 | 100.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75336-86-6
Molecular Formula C5H12N2
Molecular Weight (g/mol) 100.16
InChI Key JOMNTHCQHJPVAZ-RXMQYKEDSA-N
Synonym r---2-methylpiperazine, r-2-methylpiperazine, 2r-2-methylpiperazine, r-mp, piperazine, 2-methyl-, 2r, r-2-methyl-piperazine, 2-methylpiperazine, r, unii-p4xu643r8w
PubChem CID 7330434
IUPAC Name (2R)-2-methylpiperazine
SMILES CC1CNCCN1

Specifications

Melting Point 86.0°C to 91.0°C
Color White to Yellow
Boiling Point 155.0°C to 156.0°C
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Rotation Condition − 18.00 (20.00°C c=1,toluene)
Specific Rotation − 18.00
Solubility Information Solubility in water: soluble.
Formula Weight 100.16
Percent Purity 95%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material (R)-(-)-2-Methylpiperazine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (R)-(-)-2-Methylpiperazine
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.