missing translation for 'onlineSavingsMsg'
Learn More

(R)-2-Methyl-CBS-oxazaborolidine monohydrate, 94%, Thermo Scientific™

Product Code. 11387296
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387296 250 mg 250mg
11377296 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387296 Supplier Thermo Scientific Alfa Aesar Supplier No. L09230.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112022-83-0
Molecular Formula C18H20BNO
Molecular Weight (g/mol) 277.17
MDL Number MFCD00078440
InChI Key VMKAFJQFKBASMU-QGZVFWFLSA-N
Synonym r-2-methyl-cbs-oxazaborolidine, r-methyl oxazaborolidine, r-+-2-methyl-cbs-oxazaborolidine, r-5,5-diphenyl-2-methyl-3,4-propano-1,3,2-oxazaborolidine, r-1-methyl-3,3-diphenylhexahydropyrrolo 1,2-c 1,3,2 oxazaborole, r-me-cbs catalyst, r-3,3-diphenyl-1-methylpyrrolidino 1,2-c-1,3,2-oxazaborole, r-me cbs, r-tetrahydro-1-methyl-3,3-diphenyl-1h,3h-pyrrolo 1,2-c 1,3,2 oxazaborole, corey's catalyst
PubChem CID 9838490
IUPAC Name (3aR)-1-methyl-3,3-diphenyl-3a,4,5,6-tetrahydropyrrolo[1,2-c][1,3,2]oxazaborole
SMILES [H][C@]12CCCN1B(C)OC2(C1=CC=CC=C1)C1=CC=CC=C1

Tekniske data

Quantity 250 mg
Beilstein 9059874
Optical Rotation +74° (c=1 in Methanol)
Formula Weight 295.19 (277.17 Anhydrous)
Percent Purity 94%
Chemical Name or Material (R)-2-Methyl-CBS-oxazaborolidine monohydrate
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.