missing translation for 'onlineSavingsMsg'
Learn More

(R)-2-Methyl-CBS-oxazaborolidine, 1M soln. in toluene

Product Code. 11373596
Change view
Click to view available options
Quantity:
1 mL
5 mL
25 mL
Unit Size:
1mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11373596 25 mL 25mL
11353596 1 mL 1mL
11363596 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11373596 Supplier Thermo Scientific Alfa Aesar Supplier No. L14582.AC
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 112022-83-0 | C18H20BNO | 277.17 g/mol

(R)-2-Methyl-CBS-oxazaborolidine is used in the asymmetric reduction of prochiral ketones. Other applications include the enantioselective synthesis of α-hydroxy acids, α-amino acids, C2 symmetrical ferrocenyl diols, and propargyl alcohols. It is also used in a desymmetrizing reduction leading to (S)-4-hydroxycyclohexenone. It is useful in the production of stereospecific motifs such as α-hydroxy acids, α-amino acids, symmetrical ferrocenyl diols and propargyl alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-2-Methyl-CBS-oxazaborolidine is used in the asymmetric reduction of prochiral ketones. Other applications include the enantioselective synthesis of α-hydroxy acids, α-amino acids, C2 symmetrical ferrocenyl diols, and propargyl alcohols. It is also used in a desymmetrizing reduction leading to (S)-4-hydroxycyclohexenone. It is useful in the production of stereospecific motifs such as α-hydroxy acids, α-amino acids, symmetrical ferrocenyl diols and propargyl alcohols.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store under inert gas. It is sensitive to moisture and air. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112022-83-0
Molecular Formula C18H20BNO
Molecular Weight (g/mol) 277.17
MDL Number MFCD00078440
InChI Key VMKAFJQFKBASMU-QGZVFWFLSA-N
Synonym r-2-methyl-cbs-oxazaborolidine, r-methyl oxazaborolidine, r-+-2-methyl-cbs-oxazaborolidine, r-5,5-diphenyl-2-methyl-3,4-propano-1,3,2-oxazaborolidine, r-1-methyl-3,3-diphenylhexahydropyrrolo 1,2-c 1,3,2 oxazaborole, r-me-cbs catalyst, r-3,3-diphenyl-1-methylpyrrolidino 1,2-c-1,3,2-oxazaborole, r-me cbs, r-tetrahydro-1-methyl-3,3-diphenyl-1h,3h-pyrrolo 1,2-c 1,3,2 oxazaborole, corey's catalyst
PubChem CID 9838490
IUPAC Name (3aR)-1-methyl-3,3-diphenyl-3a,4,5,6-tetrahydropyrrolo[1,2-c][1,3,2]oxazaborole
SMILES [H][C@]12CCCN1B(C)OC2(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Density 0.925
Flash Point 4°C (39°F)
Quantity 25 mL
UN Number UN1993
Sensitivity Air and moisture sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 277.17
Concentration or Composition (by Analyte or Components) 1M soln. in toluene
Chemical Name or Material (R)-2-Methyl-CBS-oxazaborolidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.