missing translation for 'onlineSavingsMsg'
Learn More

(R)-2-Isobutylsuccinic acid-1-methyl ester, 95%, (98% ee)

Product Code. 10384782 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10384782 1 g Glass bottle 1g
10370982 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10384782 Supplier Thermo Scientific Supplier No. 363050010
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 130165-76-3 | C9H16O4 | 188.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130165-76-3
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.22
MDL Number MFCD01091032
InChI Key AJNNFMRCMKGYHA-SSDOTTSWSA-N
Synonym r-2-isobutylsuccinic acid-1-methyl ester, 3r-3-methoxycarbonyl-5-methylhexanoic acid, r-3-methoxycarbonyl-5-methylhexanoic acid, butanedioic acid,2-2-methylpropyl-, 1-methyl ester, 2r, r-2-isobutyl succinic acid 1-methyl ester, r-+-2-isobutylsuccinic acid 1-methyl ester
PubChem CID 5702633
IUPAC Name (3R)-3-methoxycarbonyl-5-methylhexanoic acid
SMILES CC(C)CC(CC(=O)O)C(=O)OC

Specifications

Color Yellow
Enantiomeric Excess 97.5% min. (GC)
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 1 g
Specific Rotation +18° to +21° (25°C, 589nm) (c=1, CH3OH)
Formula Weight 188.22
Percent Purity 95%,(98% ee)
Physical Form Viscous Liquid
Chemical Name or Material (R)-2-Isobutylsuccinic acid-1-methyl ester, 95%, (98% e.e.)
compliance-icons
Product Identifier
  • (R)-2-Isobutylsuccinic acid-1-methyl ester
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.