missing translation for 'onlineSavingsMsg'
Learn More

(R)-2-Hydroxybutanoic Acid, TRC

Product Code. 30390540
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30390540 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30390540 Supplier Toronto Research Chemicals Supplier No. TRCH977663250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2R)-2-Hydroxybutanoic Acid
CAS 20016-85-7
Molecular Formula C4 H8 O3
Quantity 250 mg
Synonym Butanoic acid, 2-hydroxy-, (2R)-, Butanoic acid, 2-hydroxy-, (R)-, Butyric acid, 2-hydroxy-, D- (8CI), (2R)-2-Hydroxybutanoic acid, (-)-2-Hydroxy-n-butyric acid, (-)-2-Hydroxybutanoic acid, (-)-2-Hydroxybutyric acid, (-)-α-Hydroxybutyric acid, (R)-2-Hydro
InChI Formula InChI=1S/C4H8O3/c1-2-3(5)4(6)7/h3,5H,2H2,1H3,(H,6,7)/t3-/m1/s1
SMILES CC[C@@H](O)C(=O)O
IUPAC Name (2R)-2-hydroxybutanoic acid
Molecular Weight (g/mol) 104.105
Formula Weight 104.047
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.