missing translation for 'onlineSavingsMsg'
Learn More

(R)-2-Hydroxy-4-phenylbutyric Acid Ethyl Ester, TRC

Product Code. 30401586
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30401586 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30401586 Supplier Toronto Research Chemicals Supplier No. TRCH949096250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 90315-82-5
Molecular Formula C12H16O3
Quantity 250 mg
Synonym (alphaR)-alpha-Hydroxybenzenebutanoic Acid Ethyl Ester, (R)-Ethyl 2-Hydroxy-4-phenylbutanoate, Ethyl (R)-alpha-Hydroxybenzenebutanoate, Ethyl 2(R)-hydroxy-4-phenylbutyrate
InChI Formula InChI=1S/C12H16O3/c1-2-15-12(14)11(13)9-8-10-6-4-3-5-7-10/h3-7,11,13H,2,8-9H2,1H3/t11-/m1/s1
SMILES CCOC(=O)[C@H](O)CCc1ccccc1
IUPAC Name ethyl (2R)-2-hydroxy-4-phenylbutanoate
Molecular Weight (g/mol) 208.25
Formula Weight 208.1099
Chemical Name or Material (R)-2-Hydroxy-4-phenylbutyric Acid Ethyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.