missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Heptyl isothiocyanate, 98%, Thermo Scientific™

Product Code. 15499119
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 737000-93-0
Density 0.91
Molecular Formula C8H15NS
Refractive Index 1.486
MDL Number MFCD05664051
Quantity 1 g
UN Number UN2922
Sensitivity Moisture sensitive
Synonym r---2-heptyl isothiocyanate, r-2-isothiocyanatoheptane, 2r-2-isothiocyanatoheptane, r---2-heptylisothiocyanate
InChI Key UPTLUDLHIIYPNC-MRVPVSSYSA-N
SMILES CCCCCC(C)N=C=S
IUPAC Name (2R)-2-isothiocyanatoheptane
Molecular Weight (g/mol) 157.275
PubChem CID 7018248
Formula Weight 157.28
Percent Purity 98%
Chemical Name or Material (R)-(-)-2-Heptyl isothiocyanate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'