missing translation for 'onlineSavingsMsg'
Learn More

(R)-2-Aminomethyl-1-Boc-pyrrolidine, 97%

Product Code. 15459147
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15459147 250 mg 250mg
15439147 1 g 1g
15449147 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15459147 Supplier Thermo Scientific Alfa Aesar Supplier No. H52419.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 259537-92-3 | C10H20N2O2 | 200.282 g/mol

Reactant in the synthesis of diamino derivatives of [1, 2, 4] triazolo [1, 5-a][1, 3, 5] triazine(potent and selective adenosine A2A receptor antagonists).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant in the synthesis of diamino derivatives of [1, 2, 4] triazolo [1, 5-a][1, 3, 5] triazine(potent and selective adenosine A2A receptor antagonists).

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 259537-92-3
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.282
MDL Number MFCD03419256
InChI Key SOGXYCNKQQJEED-MRVPVSSYSA-N
Synonym r-2-aminomethyl-1-n-boc-pyrrolidine, r-1-boc-2-aminomethyl pyrrolidine, r-tert-butyl 2-aminomethyl pyrrolidine-1-carboxylate, tert-butyl 2r-2-aminomethyl pyrrolidine-1-carboxylate, r-2-aminomethyl-1-boc-pyrrolidine, r-2-aminomethyl-pyrrolidine-1-carboxylic acid tert-butyl ester, r-2-aminomethyl-1-tert-butoxycarbonyl pyrrolidine, tert-butyl r-2-aminomethyl-1-pyrrolidinecarboxylate, r-2-aminomethyl-1-n-boc-pyyrolidine
PubChem CID 1512534
IUPAC Name tert-butyl (2R)-2-(aminomethyl)pyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC1CN

Specifications

Boiling Point 98°C to 110°C (1 mmHg)
Quantity 250 mg
UN Number UN2735
Solubility Information Slightly soluble in water.
Optical Rotation +47° (c=1 in Chloroform)
Formula Weight 200.28
Percent Purity 97%
Chemical Name or Material (R)-2-Aminomethyl-1-Boc-pyrrolidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.