missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Aminohexane, ChiPros 99+%, ee 96+%

Product Code. 11324206
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11324206 5 g 5g
11314206 1 g 1g
11334206 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11324206 Supplier Thermo Scientific Alfa Aesar Supplier No. L19304.06
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 70095-40-8 | C6H15N | 101.193 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70095-40-8
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.193
MDL Number MFCD00671627
InChI Key WGBBUURBHXLGFM-ZCFIWIBFSA-N
Synonym r-2-aminohexane, 2r-hexan-2-amine, r-2-hexylamine, 2-hexanamine, r, r-2-hexaneamine, pubchem6753, 2r-2-hexanamine, r-hexan-2-amine, 2-hexanamine, 2r, r-+-2-aminohexane
PubChem CID 6993464
IUPAC Name (2R)-hexan-2-amine
SMILES CCCCC(C)N

Specifications

Melting Point <-20°C
Density 0.755
Boiling Point 116°C
Flash Point −3°C (26°F)
Quantity 5 g
UN Number UN2733
Beilstein 1718994
Sensitivity Air Sensitive
Formula Weight 101.19
Percent Purity ≥99%
Assay ee ≥96%
Chemical Name or Material (R)-(-)-2-Aminohexane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.