missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Amino-3-methyl-1-butanol, 98%

Product Code. 10339193 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10339193 25 g Glass bottle 25g
10532252 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10339193 Supplier Thermo Scientific Acros Supplier No. 422470250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 4276-09-9 | C5H13NO | 103.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4276-09-9
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.17
MDL Number MFCD00064297
InChI Key NWYYWIJOWOLJNR-YFKPBYRVSA-N
IUPAC Name (2R)-2-amino-3-methylbutan-1-ol
SMILES CC(C)[C@@H](N)CO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 32.0°C to 36.0°C
Color White to Yellow
Density 0.9310g/mL
Boiling Point 189.0°C to 190.0°C
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Specific Gravity 0.931
Specific Rotation Condition -16.50 (20.00°C c = 10,EtOH)
Specific Rotation -16.50
Synonym 4-chloroquinaldine, quinoline, 4-chloro-2-methyl, 4-chloro-2-methyl-quinoline, 2-methyl-4-chloroquinoline, quinaldine, 4-chloro, 4-chloro-quinaldine, acmc-20a2ui, 4-chloroquinaldine hcl, quinoline,4-chloro-2-methyl
PubChem CID 77973
Formula Weight 103.16
Percent Purity 98%
Physical Form Low Melting Crystals or Crystalline Mass
Chemical Name or Material (R)-(-)-2-Amino-3-methyl-1-butanol, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.